Components
Composition:
64180 Hydroxypropyl methacrylate, 1 l
20400 2-Butoxyethanol, 100 mL
90412 Triethylene glycol dimethacrylate, 50 ml
33581 Benzoyl peroxide, 10 g
81170 Polyethylene glycol 400, 100 ml
39440 N,N-Dimethylaniline, 5 ml
Application
Used to formulate water-miscible embedding media of excellent cutting properties for microscopy
Water-miscible embedding material of excellent cutting properties for embedding of as well soft as hard tissue.
Other Notes
The components can also be ordered separately under the Fluka product numbers indicated.