|
| 20982 Fluka |
Cesium chloride Lanthanum chloride bufferfor flame atomic absorption spectrometry, buffer solution according to Schinkel |
|
|||||||
| Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |