Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
21965
Fluka
5(6)-Carboxy-X-rhodamine
BioChemika
, for fluorescence
zoom
1 of 1
Molecular Formula
C
33
H
30
N
2
O
5
Molecular Weight
534.60
CAS Number
198978-94-8
MDL number
MFCD01862979
Expand/Collapse All
Price and Availability
Descriptions
Analysis Note
partially soluble in methanol, 0.1 M Tris pH 8.0
Properties
grade
for fluorescence
product line
BioChemika
fluorescence
λ
ex
570 nm; λ
em
~595 nm in 0.1 M Tris pH 8.0
mp
≥250 °C(lit.)
Safety
Hazard Codes
Xi
Risk Statements
36/37/38
Safety Statements
26-36
WGK Germany
3
F
3-8-10
Related Categories
...
Fluorescent Probes, Labels, Particles and Stains
>
Detection
...
Functional Group Reactive Labels
>
Functional Group Reactive Labels for Immunoassays
...
Lipophilic and Membrane Probes
>
Labels and Other Fluorophores for Membrane Research
...
Nucleic Acid Stains
>
Nucleic Acid Stains For Microscopy
...
Fluorescent Labels
>
Rhodamine and Derivatives
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Related Categories
Page Options
Printer Friendly View
Ask A Scientist
Email Page
Last 5 Products Viewed
21965 (Fluka)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.