product line
BioUltra
assay
≥99.5% (T)
expl. lim.
8 %± , 65 °F
total impurities
insoluble matter, passes filter test
ign. residue
≤0.05% (as SO4)
loss
≤0.5% loss on drying, 110 °C
pH
1.0-2.0 (25 °C, 1 M in H2O)
pKa
(1) 3.13, (2) 4.76, (3) 6.4
mp
153-159 °C(lit.)
solubility
H2O: 1 M at 20 °C, clear, colorless
anion traces
chloride (Cl-): ≤5 mg/kg
oxalate (C2O42-): ≤500 mg/kg
phosphate (PO4-): ≤5 mg/kg
sulfate (SO42-): ≤20 mg/kg
tartrate (C4H4O62-): ≤2000 mg/kg
cation traces
Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg
λ
1 M in H2O
UV absorption
λ: 260 nm Amax: 0.20
λ: 280 nm Amax: 0.10
Gene Information