Analysis Note
λmax. ∼610 nm; E(1%,1 cm) ∼500-700 (H2O)
Biochem/physiol Actions
P2X7 purinergic receptor antagonist.
Other Notes
Protein stain for gel electrophoresis. Does not penetrate the gel and is three times more sensitive than Naphthol Blue Black B1; Simple and sensitive protein assay2; Rapid, sensitive and versatile assay for proteins; free amino acids and several chemicals that interfere with the Lowry procedure cause no interference3.
Legal Information
Coomassie is a registered trademark of Imperial Chemical Industries Ltd.
® Registered Trademark of Imperial Industries PLC