Kit component also available separately
Polyethylene glycol 1500 solution (Fluka 73034) 10 mL
73034
Polyethylene glycol 200 (Fluka 88440) 10 mL
88440
Polyethylene glycol 20,000 solution (Fluka 87006) 10 mL
87006
Polyethylene glycol 3,350 solution (Fluka 83272) 10 mL
83272
Polyethylene glycol 500 dimethyl ether (Fluka 81313) 10 mL
Polyethylene glycol 600 (Fluka 87333) 10 mL
87333
Polyethylene glycol 6,000 solution (Fluka 81304) 10 mL
81304
Polyethylene glycol 8,000 solution (Fluka 83271) 10 mL
83271
Polyethylene glycol 2,000 monomethyl ether solution (Fluka 83918) 10 mL
Polyethylene glycol 5,000 monomethyl ether solution (Fluka 90364) 10 mL
90364
Polyethylene glycol 550 monomethyl ether solution (Fluka 71578) 10 mL
71578
Polypropylene glycol P 1,200 (Fluka 81370) 10 mL
81370
Polypropylene glycol P 2,000 (Fluka 81380) 10 mL
81380
Polypropylene glycol P 400 (Fluka 81350) 10 mL
81350
Silicone antifoam, emulsion, 30% in water (Fluka 85390) 10 mL
85390
Silicone oil DC 200 (Fluka 85412) 10 mL
85412
Silicone oil DC 710 (Fluka 85427) 10 mL
85427
Triethylene glycol monobutyl ether (Fluka 90440) 10 mL
90440
Triethylene glycol dimethyl ether (Fluka 90420) 10 mL
90420
Brij
® 96 V (Fluka 16011) 10 mL
16011