product line
BioUltra
assay
≥98.0% (RT)
total impurities
insoluble matter, passes filter test
pH
3.5-5.0 (25 °C, 1 M in H2O)
mp
67-71 °C
solubility
H2O: 1 m at 20 °C, clear, colorless
anion traces
sulfate (SO42-): ≤50 mg/kg
cation traces
Al: ≤5 mg/kg
As: ≤0.5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg
abs.
absorption1M/270 nm, H2O cut-off
λ
1 M in H2O
UV absorption
λ: 280 nm Amax: 0.3
storage temp.
2-8°C