product line
BioUltra
assay
≥99.0% (RT)
optical activity
[α]20/D +5.6±0.3°, c = 5% in 5 M HCl
total impurities
insoluble matter, passes filter test
≤0.5% foreign amino acids
ign. residue
≤0.05% (as SO4)
solubility
H2O: 1 M at 20 °C, clear, colorless
anion traces
sulfate (SO42-): ≤100 mg/kg
cation traces
Al: ≤5 mg/kg
As: ≤0.5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
NH4+: ≤500 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg
λ
1 M in H2O
UV absorption
λ: 260 nm Amax: 1.0
λ: 280 nm Amax: 0.3