| 33150 Fluka |
Sodium diphenylamine-4-sulfonateindicator for the determination of the redox potential, ACS reagent, E0 in sulfuric acid 1 mol/l = +0,83 volt, rH = 27−29 |
|
|||||||||
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |