General description
Immobilized rCBD-Protein L on cellulose beads (covalently cross-linked). Designed for quick and efficient purification of immunoglobulins (Ig) and Ig fragments (Fab and scFr). ~2.5 mg rCBD-Protein L per mL cellulose suspension.
Other Notes
An immobilized rCBD-Protein L matrix (CBD: Cellulose Binding Proteins) immobilized on cellulose beads. Direct purification of monoclonal antibodies from media supplemented with FCS1
Legal Information
CBinD is a trademark of CBD Technologies, Inc.