Features and Benefits
Each strip has an identification scale and printed pH scale. The strips can be utilized in colored solutions and suspensions with very high precision.
Packaging
Packages of 200 measuring strips.
Other Notes
This former Riedel-de Haën product is now being offered as Fluka brand. During the transition, you still may receive the Riedel product. The rebranding has no impact on product performance or quality. For further information see
www.sigma-aldrich.com/rebranding. Printing link to product
rebranding statement.