Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
44617
Fluka
6-Carboxy-X-rhodamine
N
-succinimidyl ester
BioChemika
zoom
1 of 1
Molecular Formula
C
37
H
33
N
3
O
7
Molecular Weight
631.67
CAS Number
216699-36-4
MDL number
MFCD00467156
Expand/Collapse All
Price and Availability
Descriptions
Other Notes
Real-time PCR for nucleic acid detection using labeled primers or nucleotides with this rhodamine X derivative
Properties
product line
BioChemika
suitability
in accordance for fluorescence
storage temp.
−20°C
Safety
Hazard Codes
Xi
Risk Statements
36/37/38
Safety Statements
26-36
WGK Germany
3
F
8-10-21
Related Categories
...
Fluorescent Probes, Labels, Particles and Stains
>
Detection
...
Nucleic Acid Stains
>
Nucleic Acid Stains For Microscopy
...
Fluorescent Labels
>
Rhodamine and Derivatives
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Related Categories
Page Options
Printer Friendly View
Ask A Scientist
Email Page
Last 5 Products Viewed
44617 (Fluka)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.