| 46180 Fluka |
EWN solventStandard Fluka, tested for the determination of the viscosity value of cellulose acc. to ISO 5351 |
|
|||||||||
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |