Biochem/physiol Actions
Mode of Action: Inhibits bacterial cell wall synthesis.
Antimicrobial spectrum: Gram-positive bacteria
Legal Information
VETRANAL is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.
Other Notes
This former Riedel-de Haën product is now being offered as Fluka brand. During the transition, you still may receive the Riedel product. The rebranding has no impact on product performance or quality. For further information see
www.sigma-aldrich.com/rebranding. Printing link to product
rebranding statement.