Features and Benefits
EVOblue® 30 is a fluorescent dye for covalently labeling a wide range of biological molecules. The lower excitation energy needed for EVOblue 30 causes less photodestruction and exceeds the lifetime of the fluorescent signal compared to common dyes. EVOblue 30 is optimized for an excitation at 635 nm and an emission around 670 nm - both allowing the use of common laser and filter sets. In contrast to commercially available red excitable dyes, EVOblue 30 reveals a high chemical stability and suitablity for high performance fluorescence applications.
Other Notes
Fluorescent marker for the red spectral region which is very stable1
Legal Information
EVOblue is a trademark of Evotec Technologies GmbH.