grade
for luminescence
product line
BioUltra
assay
≥99.0% (NT)
total impurities
insoluble matter, passes filter test
ign. residue
≤0.05% (as SO4)
loss
≤0.05% loss on drying, 20 °C (HV)
fluorescence
λex 200-600; λem 300-800 nm (corresponds)
pKa (25 °C)
2.35
mp
240 °C (dec.)(lit.)
solubility
1 M HCl: 1 M at 20 °C, clear, colorless
anion traces
chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg
cation traces
Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
NH4+: ≤200 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg
λ
1 M in 1 M HCl
UV absorption
λ: 260 nm Amax: 0.01
λ: 280 nm Amax: 0.01