A source of protein and other important growth factors for microorganisms like Clostridia, Bacillus cereus, Staphylococci and Micrococci. One kg of egg powder and 3 liters of water is equivalent to the original egg liquid; 1 kg egg powder is equivalent to 80 eggs.