Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
60610
Fluka
Canada balsam
BioChemika
, for microscopy
Synonym
Balsam Canada
CAS Number
8007-47-4
MDL number
MFCD00132800
EG/EC Number
232-362-2
Expand/Collapse All
Price and Availability
Properties
grade
for microscopy
product line
BioChemika
refractive index
n
20/D
1.522(lit.)
n
20/D
1.522
viscosity
2000-4500 mPa.s (25 °C)
density
0.99 g/mL at 20 °C
0.99 g/mL at 25 °C(lit.)
References
reference
FT-IR
2
(1), 298:D /
FT-IR
2
(1), 298:A
Safety
WGK Germany
2
RTECS
CP2352500
Flash Point(F)
144 °F
Flash Point(C)
62 °C
Related Categories
...
Microscopy Reagents
>
Immersion Oils
...
Hematology and Histology
>
Routine Histology Stains
Scroll Up
Scroll Down
Related Information
FT-IR Raman
MSDS
Certificate of Analysis
More Information
Links
Similar Products
Related Categories
Page Options
Printer Friendly View
Ask A Scientist
Email Page
Last 5 Products Viewed
60610 (Fluka)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.