Biochem/physiol Actions
Activates the decomposition of hydrogen peroxide, Natural antioxidant used to study roles of reactive oxygen species in gene expression and apoptosis.
Other Notes
Sales restrictions may apply
Structure1,2,3; Properties4,5; Used for analytical purposes, e.g. the determination of cysteamine6; Hydrogen peroxide detection from activated phagocytes by a peroxidase-independent method7;
Unit Definition
1 U corresponds to the amount of enzyme which decomposes 1 μmol H2O2 per minute at pH 7.0 and 25°C