Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
61764
Fluka
Lectin from
Phaseolus vulgaris
BioChemika
, lyophilized, powder
Synonym
PHA
Red kidney bean Agglutinin
MDL number
MFCD00165256
EG/EC Number
232-718-7
Expand/Collapse All
Price and Availability
Properties
product line
BioChemika
form
powder
quality
lyophilized
mol wt
M
r
~128000(lit.)
M
r
~128000
storage temp.
2-8°C
Safety
Safety Statements
22-24/25
RTECS
TJ3470000
F
10-21
Related Categories
...
Lectins
>
Phaseolus vulgaris
(Red kidney bean)
...
Biochemicals and Reagents
>
Lectins
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Similar Products
Related Categories
Page Options
Printer Friendly View
Ask A Scientist
Email Page
Last 5 Products Viewed
61764 (Fluka)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.