Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Product No. Only)
Supplier X Reference
Substructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
62107
Fluka
Lewatit
®
MonoPlus TP 214
ion exchange resin, macroporous
CAS Number
109945-55-3
MDL number
MFCD01867339
Expand/Collapse All
Price and Availability
Descriptions
Application
Chelating resin especially suited for the removal of metals in hydrometallurgical processes and for the extraction/recycling of mercury and silver.
Legal Information
Lewatit is a registered trademark of Lanxess Deutschland GmbH.
MonoPlus is a trademark of Bayer AG.
Properties
form
macroporous
loss
40-50% loss on drying, 110 °C
matrix
styrene-divinylbenzene (macroporous)
matrix active group
thiourea functional group
particle size
0.4-1.25 mm
400-1250 μm
Safety
Safety Statements
24/25
WGK Germany
3
Flash Point(F)
482 °F
Flash Point(C)
250 °C
Related Categories
...
Resins & LC Separation Media
>
Chelating Ion Exchange Media
...
Cation
>
Lewatit
®
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Last 5 Products Viewed
62107 (Fluka)
en-us-footer US
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.