Features and Benefits
Partially purified, soluble extracts containing FMN-dependent luciferase and NADH- and NADPH-dependent FMN reductases. Produces light in a system containing FMN, NADH or NADPH, and n-decyl aldehyde.
Other Notes
Produces light in a system with FMN, NADH and an aliphatic aldehyde containing more than 8-C atoms.; The pack size is sufficient for more than 200 determinations of NADH in a concentration range of 10-10 mol/l.; A solution in water (5 mg lyophilisate/0.5 ml) is stable for at least one week and deep-frozen for several months.; The biochemistry and molecular biology of bacterial bioluminescence1,2