Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
62660
Fluka
LR Accelerator
MDL number
MFCD00217542
Expand/Collapse All
Price and Availability
Properties
total impurities
≥20% N,N-dimethyl-p-toluidine
Safety
Hazard Codes
T
Risk Statements
23/24/25-33-52/53
Safety Statements
28-36/37-45-60
RIDADR
UN 1708 6.1/PG 2
WGK Germany
3
Flash Point(F)
248 °F
Flash Point(C)
120 °C
F
8-10-23
Related Categories
...
Embedding and Mounting Media
>
Embedding Media
...
Hematology and Histology
>
Routine Histology Stains
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Related Categories
Page Options
Printer Friendly View
Ask A Scientist
Email Page
Last 5 Products Viewed
62660 (Fluka)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.