Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
70161
Fluka
Yeast Extract
BioChemika
, for microbiology
MDL number
MFCD00132599
EG/EC Number
232-387-9
Expand/Collapse All
Price and Availability
Descriptions
Application
A water soluble extract of autolyzed yeast cells. Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates and can be used as additive for culture media.
Properties
grade
for microbiology
product line
BioChemika
total impurities
~11% total nitrogen (N)
~5% amino nitrogen
ign. residue
≤15%
loss
≤6% loss on drying
pH
7.0±0.2 (2% in H
2
O)
solubility
H
2
O: 2%, clear, yellow
Safety
WGK Germany
3
RTECS
ZF6610000
F
3
Related Categories
...
Media for Water Control
>
Media for Water Control (General)
...
Base Ingredients
>
Yeast Extracts
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Similar Products
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Email Page
Last 5 Products Viewed
70161 (Fluka)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.