grade
for luminescence
product line
BioUltra
assay
≥99.0% (NT)
form
solid
autoignition temp.
1112 °F
total impurities
insoluble matter, passes filter test
loss
≤0.5% loss on drying, 150 °C
pH
8.5-10.0 (25 °C, 3 M in H2O)
pKa
4.76 (acetic acid)
mp
>300 °C (dec.)(lit.)
solubility
H2O: 3 M at 20 °C, clear, colorless
density
1.528 g/mL at 25 °C(lit.)
anion traces
chloride (Cl-): ≤20 mg/kg
phosphate (PO4-): ≤5 mg/kg
sulfate (SO42-): ≤20 mg/kg
cation traces
Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤10 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤10 mg/kg
Zn: ≤5 mg/kg
λ
3 M in H2O
UV absorption
λ: 260 nm Amax: 0.03
λ: 280 nm Amax: 0.02
suitability
in accordance for luminescence test