Components
Cocktail Composition
1.50 wt% Tridodecylmethylammonium nitrate (91664)
16.25 wt% 2-Nitrophenyl octyl ether (73732)
1.93 wt% Nitrocellulose, 35% in isopropanol
0.25 wt% Methyltriphenylphosphonium bromide (130079)
5.75 wt% Poly(vinyl chloride) high molecular weight (81392)
74.32 wt% Tetrahydrofuran (87369)
Other Notes
Nitrate-selective membrane solution for the determination of intracellular NO3- with microelectrodes1,2
Legal Information
Selectophore is a registered trademark of Sigma-Aldrich GmbH.