Kit component also available separately
N-(2-Acetamido)iminodiacetic acid pH 6.5; 0.1M – Ammonium phosphate monobasic 1.0M solution
93751
Ammonium phosphate dibasisch 0.1M – TRIS hydrochloride pH 8.5; 0.1M – Sodium phosphate dibasisch 0.5M – Kalium phosphate dibasisch 0.5M solution
80429
Ammonium phosphate dibasic 0.1M – TRIS hydrochloride pH 8.5; 0.1M – Polyethylene glycol 6000 12% (w/v) solution
90173
Ammonium sulfate 0.1M – HEPES-Na pH 7.5; 0.1M – Sodium phosphate dibasisch 0.5M – Kalium phosphate dibasisch 0.5M solution
86952
Ammonium sulfate 0.1M – HEPES-Na pH 7.5; 0.1M – Polyethylene glycol 400 18% (v/v) solution
85901
Ammonium sulfate 0.1M – HEPES-Na pH 7.5; 0.1M – Polyethylene glycol 4000 10% (w/v) solution,
BioChemika 76565
Ammonium sulfate 0.2M – Sodium acetate pH 4.6; 0.1M – Polyethylene glycol 4000 10% (w/v) solution
80361
Magnesiumchloride 0.1M – Sodium citrate tribasic pH 5.6; 0.1M – 2-Methyl-2,4-pentanediol 4% (v/v) solution,
BioChemika 71236
Magnesiumchloride 0.1M – Sodium citrate tribasic pH 5.6; 0.1M – 2-Methyl-2,4-pentanediol 4% (v/v) solution,
BioChemika 71236
Sodium chloride 0.15M – TRIS hydrochloride pH 8.0; 0.1M – Polyethylene glycol 6000 12 % (w/v) solution,
BioChemika 72989
Sodium chloride 0.1M – Sodium citrate tribasic pH 5.6; 0.1M – 2-Methyl-2,4-pentanediol 12% solution,
BioChemika 73347
Zinc acetate 0.1M – Sodium acetate pH 4.6; 0.1M – Polyethylene glycol 4000 12 % (v/v) solution,
BioChemika 74102
Lithium sulfate 0.1M – Sodium citrate tribasic pH 5.6; 0.1M – Polyethylene glycol 4000 12 % (w/v) solution,
BioChemika 76527
Sodium citrate tribasic 0.1M – TRIS hydrochloride pH 8.5; 0.1M – Polyethylene glycol 400 5% (v/v) solution,
BioChemika 77484
TRIS hydrochloride pH 8.5; 0.1M – Ammonium sulfate 0.5M solution,
BioChemika 77927
N-(2-Acetamido)iminodiacetic acid pH 6.5; 0.1M – 2-Methyl-2,4-pentanediol 12% (v/v) solution
79971
Lithium sulfate 0.1M – N-(2-Acetamido)iminodiacetic acid pH 6.5; 0.1M – Magnesium sulfate 1.0M solution
80059
Sodium chloride 0.1 M – Sodium acetate pH 4.6; 0.1M – 2-Methyl-2,4-pentanediol 12% (w/v) solution
80070
Sodium citrate tribasic pH 5.6; 0.1M – 2-Propanol 10% (v/v) solution
80088
Sodium chloride 0.1M – TRIS hydrochloride pH 8.0; 0.1M – 2-Methyl-2,4-pentanediol 12% (v/v) solution
80092
Lithium sulfate 0.1M – TRIS hydrochloride pH 8.5; 0.1M – 2-Methyl-2,4-pentanediol 12% (v/v) solution
80428
Lithium sulfate 0.1M – HEPES-Na pH 7.5; 0.1M – Potassium sodium tartrate 0.1M solution
80566
Lithium sulfate 0.1M – Sodium citrate tribasic pH 5.6; 0.1M – Polyethylene glycol 6000 12% (w/v) solution
80807
Sodium acetate pH 4.6; 0.1M – Polyethylene glycol 4000 12% (w/v) solution
80877
Potassium sodium tartrate 0.1M – TRIS hydrochloride pH 8.5; 0.1M – Magnesium sulfate 0.4M solution
81855
Sodium chloride 0.1M – Sodium acetate pH 4.6; 0.1M – 2-Propanol 12% (v/v) solution
82178
Sodium citrate tribasic pH 5.6; 0.1M – Magnesium sulfate 1.0M solution
82767
Sodium acetate pH 4.6; 0.1M – Magnesium sulfate 1.0M solution
82851
Sodium chloride 0.1M – Sodium citrate tribasic pH 5.6; 0.1M – Polyethylene glycol 4000 12% (w/v) solution
85242
Magnesiumchloride 0.1M – HEPES-Na pH 7.5; 0.1M – Polyethylene glycol 400 18% (v/v) solution
85852
Magnesium chloride 0.1M – Sodium acetate pH 4.6; 0.1M – Polyethylene glycol 400 18% (v/v) solution
86303
TRIS hydrochloride pH 8.5; 0.1M – Sodium chloride 0.1M solution
86782
Magnesium chloride 0.1M – N-(2-Acetamido)iminodiacetic acid pH 6.5; 0.1M – Polyethylene glycol 6000 12% (w/v) solution
86974
TRIS hydrochloride pH 8.5; 0.1M – Lithium sulfate 0.2M solution
87772
TRIS hydrochloride pH 8.5; 0.1M – Sodium acetate 0.1M solution
88411
Lithium sulfate 0.1M – N-(2-Acetamido)iminodiacetic acid pH 6.5; 0.1M – Polyethylene glycol 4000 12% (w/v) – 2-Propanol 2 % (v/v) solution
89468
Sodium chloride 0.1M – HEPES-Na pH 7.5; 0.1M – Polyethylene glycol 4000 10% (w/v) solution
89729
Sodium citrate tribasic 0.1M – HEPES-Na pH 7.5; 0.1M – 2-Methyl-2,4-pentanediol 12%(v/v) solution
89805
Sodium citrate tribasic pH 5.6; 0.1M – Sodium chloride 0.1M solution
91150
Sodium acetate pH 4.6; 0.1M – Ammonium sulfate 1.0M solution
92189
Magnesium sulfate 0.6M – HEPES-Na pH 7.5; 0.1M – 2-Methyl-2,4-pentanediol 4% (v/v) solution
92635
HEPES-Na pH 7.5; 0.1M – Sodium citrate tribasic 1.0M solution
92684
Lithium sulfate 0.1M – Sodium citrate tribasic pH 5.6; 0.1M – Polyethylene glycol 400 4% (v/v) solution
94293
Magnesium chloride 0.1M – Sodium acetate pH 4.6; 0.1M – Polyethylene glycol 6000 12% (w/v) solution
94588
Lithium sulfate 0.1M – Sodium acetate pH 4.6; 0.1M – Ammonium phosphate monobasic 1.0M solution
94783
Lithium sulfate 0.3M – N-(2-Acetamido)iminodiacetic acid pH 6.5; 0.1M – Polyethylene glycol 400 4% (v/v) solution
95773
Sodium chloride 0.1M – Sodium acetate pH 4.6; 0.1M – Polyethylene glycol 6000 12% (w/v) solution
96904
N-(2-Acetamido)iminodiacetic acid pH 6.5; 0.1M – Ammonium sulfate 1.0M solution,
BioChemika 72639
HEPES-Na pH 7.5; 0.1M – Potassium sodium tartrate 1.0M solution
79049