Kit component also available separately
α-1,3-Galactosyl-Transferase recombinant from
E. coli,
BioChemika, solution (clear, colorless), ~0.5 U/mL
77038
Kit component only
SN-GAL-1
Kit component also available separately
Manganese(II) chloride tetrahydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)
63536
Kit component only
Trizma® Buffer substance, BioChemika, pH 7.0
Albumin from bovine serum, BioChemika, lyophilized, crystallized, ≥98.0% (GE)
Phosphatase, Alkaline from bovine intestinal mucosa, BioChemika, solution (clear), >10000 U/mL