Kit component also available separately
Imidazole pH 6.5; 0.07 M - Sodium acetate 0.7 M - Glycerol 30% solution,
BioChemika 71591
Sodium citrate tribasic pH 5.6; 0.07 M – Ammonium phosphate monobasic 0.7 M – Glycerol 30% (v/v) solution,
BioChemika 72871
Magnesium chloride 0.18 M – HEPES-Na pH 7.5; 0.09M – 2-Propanol 27% (w/v) – Glycerol 10% solution,
BioChemika 72872
Sodium acetate 0.17 M – TRIS hydrochloride pH 8.5; 0.085 M – Polyethylene glycol 4000 25.5% (w/v) – Glycerol 15% (v/v) solution,
BioChemika 73803
Ammonium acetate 0.16M – TRIS hydrochloride pH 8.5; 0.08M – 2-Propanol 24% (v/v) – Glycerol 20% (v/v) solution,
BioChemika 74613
Sodium citrate tribasic 0.14M – Sodium cacodylate pH 6.5; 0.07M – 2-Propanol 21% (v/v) – Glycerol 30% (v/v) solution,
BioChemika 74614
Magnesium chloride 0.2M – HEPES Na pH 7.5; 0.1M – Polyethylene glycol 400 30% (w/v) solution,
BioChemika 76525
Ammonium acetate 0.2M – Sodium citrate tribasic pH 5.6; 0.1M – 2-Methyl-2,4-pentanediol 30% (w/v) solution,
BioChemika 76658
HEPES Na pH 7.5; 0.065M – Potassium sodium tartrate 0.52M – Glycerol 35% (v/v) solution,
BioChemika 76659
Ammonium acetate 0.17M – Sodium citrate tribasic pH 5.6; 0.085M – Polyethylene glycol 4000 25.5% (w/v) – Glycerol 15% (v/v) solution,
BioChemika 77437
Ammonium acetate 0.17M – Sodium acetate pH 4.6; 0.085M – Polyethylene glycol 4000 25.5% (w/v) – Glycerol 15% (v/v) solution,
BioChemika 77582
Magnesium acetate 0.2M – Sodium cacodylate pH 6.5; 0.1M – 2-Methyl-2,4-pentanediol 30% (v/v) solution
79974
Sodium citrate tribasic 0.2M – TRIS hydrochloride pH 8.5; 0.1M – Polyethylene glycol 400 30% (w/v) solution
80055
Ammonium sulfate 0.17M – Sodium cacodylate pH 6.5; 0.085M – Polyethylene glycol 8000 25.5% (w/v) – Glycerol 15% (v/v) solution
80074
Lithium sulfate 0.17 M – TRIS hydrochloride pH 8.5; 0.085M – Polyethylen glycol 4000 25.5% (w/v) – Glycerol 15% (v/v) solution
80091
Magnesium acetate 0.16 M – Sodium cacodylate pH 6.5; 0.08M – Polyethylene glycol 8000 16% (w/v) – Glycerol 20% solution
80093
Sodium citrate tribasic 0.14M, HEPES Na pH 7.5; 0.07M, 2-Propanol 14% (v/v), Glycerol 30% solution
80384
Sodium acetate 0.17M – Sodium cacodylate pH 6.5; 0.085M – Polyethylene glycol 8000 25.5% (w/v) – Glycerol 15% (v/v) solution
80387
Calcium chloride 0.19 M – HEPES-Na pH 7.5; 0.095M – Polyethylene glycol 400; 26.6% (w/v) – Glycerol 5% solution
80556
Ammonium sulfate 0.17 M – Polyethylene glycol 8000 25.5% (w/v) – Glycerol 15% solution
80803
HEPES-Na pH 7.5; 0.085M – Polyethylene glycol 400 1.7% (v/v) – Ammonium sulfate 1.7M – Glycerol 15% (v/v) solution
80864
HEPES-Na pH 7.5; 0.075M – Sodium phosphate monobasic 0.6M – Potassium phosphate monobasic 0.6M – Glycerol 25% (v/v) solution
80924
Lithium sulfate 0.8 M – Polyethylene glycol 8000 1.6% (w/v) – Glycerol 20% (v/v) solution
81696
Lithium sulfate 0.4 M – Polyethylene glycol 8000 12% (w/v) – Glycerol 20% (v/v) solution
81713
Sodium formate 3.6 M – Glycerol 10% solution
81892
Sodium acetate pH 4.6; 0.07M – 1.4 M Sodium formate – Glycerol 30% (v/v) solution
81893
Sodium acetate pH 4.6; 0.07M – Polyethylene glycol 4000 5.6% (w/v) – Glycerol 30% solution
82216
HEPES-Na pH 7.5; 0.085M – 2-Propanol 8.5% (v/v) – Polyethylene glycol 4000 17% (w/v) – Glycerol 15% (v/v) solution
82809
Sodium acetate pH 4.6; 0.08 M – Ammonium sulfate 1.6 M – Glycerol 20% (v/v) solution
83196
Zinc acetate 0.16M – Sodium cacodylate pH 6.5; 0.08M – Polyethylene glycol 8000 14.4% (w/v) – Glycerol 20% (v/v)
83198
Potassium phosphate monobasic 0.04M – Polyethylene glycol 8000 16% (w/v) – Glycerol 20% (v/v) solution
83617
Calcium acetate 0.16 M – Sodium cacodylate pH 6.5; 0.08M – Polyethylene glycol 8000 14.4% (w/v) – Glycerol 20%
84461
TRIS hydrochloride pH 8.5; 0.08 M – Ammonium phosphate monobasic 1.6 M – Glycerol 20% (v/v) solution
Ammonium sulfate 0.17 M – Polyethylene glycol 4000 25.5% (w/v) – Glycerol 15% solution
87429
Ammonium sulfate 1.5 M – Glycerol 25% solution
89252
TRIS hydrochloride pH 8.5; 0.065M – Polyethylene glycol 8000 5.2% (w/v) – Glycerol 35% (v/v) solution
89725
HEPES-Na pH 7.5; 0.09M – Sodium citrate tribasic 1.26M – Glycerol 10% solution
89734
Polyethylene glycol 1500 24% (w/v) – Glycerol 20% (v/v) solution
90213
Magnesium formate 0.1M – Glycerol 50% (v/v) solution
91638
Sodium citrate tribasic pH 5.6; 0.095M – 2-Propanol 19% (v/v) – Polyethylene glycol 4000 19% (w/v) – Glycerol 5% solution
91959
Calcium chloride 0.02 M – Sodium acetate 0.1M – 2-Methyl-2,4-pentanediol 30% (v/v) solution
92599
Magnesium chloride 0.16M – TRIS hydrochloride pH 8.5; 0.08M – Polyethylene glycol 4000 24% (w/v) – Glycerol 20% (v/v) solution
93189
Sodium citrate tribasic 0.2M – HEPES-Na pH 7.5; 0.1M – 2-Methyl-2,4-pentanediol 30% (v/v) solution
93949
TRIS hydrochloride pH 8.5; 0.075 M – Ammonium sulfate 1.5 M – Glycerol 25% (v/v) solution
94030
Ammonium phosphate monobasic 0.26 M – Glycerol 35% (v/v) solution
96347
Potassium sodium tartrate 0.26 M – Glycerol 35% (v/v) solution
96864
Calcium chloride 0.14M – Sodium acetate pH 4.6; 0.07M – 2-Propanol 14% (v/v) – Glycerol 30% (v/v) solution,
BioChemika 73806
Sodium cacodylate pH 6.5; 0.07M – Sodium acetate 0.98M – Glycerol 30% (v/v) solution
79973
Ammonium sulfate 0.16 M – Sodium acetate pH 4.6; 0.08M – Polyethylene glycol 4000 20% (w/v) – Glycerol 20% (v/v) solution
80053
HEPES Na pH 7.5; 0.075M – Lithium sulfate 1.125M – Glycerol 25% (v/v) solution
80084