Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
76259
Fluka
Paraplast
®
X-tra
BioChemika
, for electron microscopy
Expand/Collapse All
Price and Availability
Descriptions
Application
embedding medium from a unique blend of highly purified paraffin and low MW-polymers for lower infiltration and better compression resistance
Legal Information
Paraplast is a registered trademark of McCormick Scientific.
Properties
grade
for electron microscopy
product line
BioChemika
thickness
≤2 μm , sectioning: continuous ribbons
mp
50-54 °C(lit.)
storage temp.
2-8°C
Safety
Risk Statements
22-36/37/38
Safety Statements
26
Related Categories
...
Embedding and Mounting Media
>
Embedding Media
...
Hematology and Histology
>
Routine Histology Stains
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Email Page
Last 5 Products Viewed
76259 (Fluka)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.