Frequently Asked Questions
Other Notes
Luminescent oxygen probe in optic sensors1,2; Studies on oxygen in skin3,4
Features and Benefits
Ruthenium-tris(4,7-diphenyl-1,10-phenanthroline) dichloride (Ru(dpp)) and Tris(4,7-diphenyl-1,10-phenanthroline)-ruthenium hexafluorophosphate (Ru(dpp)3(PF6)2) have been widely used as probes for luminescent detection and quantitation of oxygen.
The fluorescence of both dyes (abs. λmax 455 nm, luminescence λmax 613 nm) is strongly reduced by molecular oxygen due to dynamic quenching. Therefore, it is a viable oxygen probe based on either measurement of intensity or decay time.
Ruthenium-tris(4,7-diphenyl-1,10-phenanthroline) dichloride has been used for (fiber) optic sensors, studies on oxygen in skin and skin tumors, measurement of oxygen flux through skin and for oxygen imaging.