Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
77275
Fluka
Perfluorokerosene
for mass spectroscopy, high boiling
zoom
1 of 1
MDL number
MFCD00132835
Expand/Collapse All
Price and Availability
Properties
grade
for mass spectroscopy
refractive index
n
20/D
1.330
bp
210-260 °C
>300 °C(lit.)
Safety
Safety Statements
23-24/25
WGK Germany
3
Related Categories
...
Mass Spectrometry
>
Matrices for MALDI Analysis
...
Mass Spectrometry (MS) & LC-MS
>
Reagents for Mass Spectrometry (MS)
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Similar Products
Related Categories
Page Options
Printer Friendly View
Ask A Scientist
Email Page
Last 5 Products Viewed
77275 (Fluka)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.