product line
BioUltra
concentration
1.5 M in H2O (colorless solution at 20 °C)
total impurities
insoluble matter, passes filter test
pH
7.0-8.0 (25 °C, 1.5 M in H2O)
anion traces
chloride (Cl-): ≤1 mg/kg
sulfate (SO42-): ≤1 mg/kg
cation traces
Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/L
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg
λ
1.5 M in H2O
UV absorption
λ: 260 nm Amax: 0.070
λ: 280 nm Amax: 0.040