Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
79807
Fluka
Micro particles based on polystyrene
size: 25 μm
Synonym
Latex beads from PS
MDL number
MFCD00243243
Expand/Collapse All
Price and Availability
Descriptions
Analysis Note
For every lot exact values of particle size and standard deviation are determined with an accuracy of 0.01 μm using a Coulter Multisizer.
Properties
form
suspension (aqueous)
crosslinking
2 % cross-linked
concentration
10% (solids)
particle size
25 μm std dev <0.4 μm, coeff var <2%
density
1.05 g/cm
3
storage temp.
2-8°C
Safety
WGK Germany
3
Flash Point(F)
410 °F
Flash Point(C)
210 °C
Related Categories
...
M
>
METI - MZ
...
Microparticles
>
Microparticles based on polystyrene
...
Reference/Calibration Standards
>
Particle Size Standards
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Similar Products
Related Categories
Page Options
Printer Friendly View
Ask A Scientist
Email Page
Last 5 Products Viewed
79807 (Fluka)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.