Kit component also available separately
HEPES pH 7.5; 0.05 M − Magnesium chloride 80 mM − Spermine 2.5 mM solution,
BioChemika 28674
Cacodylate pH 6.0; 0.05 M − Magnesium chloride 18 mM − Spermine 2.25 mM − Copper sulfate 1 mM − Isopropanol 9% solution,
BioChemika 76475
Cacodylate pH 6.5; 0.05 M − Magnesium chloride 18 mM − Spermine 0.9 mM − Cobalt(III)-hexamine 1.8 mM − Isopropanol 9% solution,
BioChemika 42393
Cacodylate pH 6.5; 0.05 M − Magnesium chloride 18 mM − Spermine 2.25 mM − Isopropanol 9% solution,
BioChemika 89253
Cacodylate pH 7.0; 0.05 M − Magnesium chloride 18 mM − Spermine 2.25 mM − Cobalt(III)-hexamine 0.9 mM − 2-Methyl-2,4-pentanediol 4.5% solution,
BioChemika 67516
Cacodylate pH 6.5; 0.05 M − Magnesium chloride 36mM − Spermine 2.25 mM − PEG 400 5% solution,
BioChemika 90746
Sodium succinate pH 5.5; 0.05 M − Magnesium chloride 10 mM − Cobalt(III)-hexamine 2.0 mM − Isopropanol 10% solution,
BioChemika 89264
Cacodylate pH 6.0; 0.05 M − Magnesium chloride 20 mM − Spermine 1.0 mM − Ethanol 15% solution,
BioChemika 90971
Cacodylate pH 7.0; 0.05 M − Magnesium chloride 20 mM − Spermine 1.0 mM − Cobalt(III)-hexamine 1.0 mM − Ethanol 15% solution,
BioChemika 51189
Cacodylate pH 7.0; 0.05 M − Magnesium chloride 5 mM − Spermidine 1.0 mM −
tert-Butanol 10% solution,
BioChemika 19693
Cacodylate pH 7.0; 0.05 M − Magnesium chloride 30 mM − Spermine 2.5 mM − PEG 400 5% solution,
BioChemika 93084
Cacodylate pH 6.5; 0.05 M − Magnesium chloride 100 mM − Cobalt(III)-hexamine 2.0 mM − Isopropanol 5% solution,
BioChemika 30517
TRIS pH 8.0; 0.05 M − Magnesium chloride 10 mM − Cobalt(III)-hexamine 1.0 mM − Ethanol 20% solution,
BioChemika 51804
HEPES pH 7.5; 0.05 M − Magnesium chloride 20 mM − Spermine 1.0 mM − PEG 8000 5% solution,
BioChemika 90748
Cacodylate pH 6.0; 0.05 M − Magnesium chloride 20 mM − Spermine 2.5 mM − PEG 4000 5% solution,
BioChemika 95109
Cacodylate pH 6.0; 0.05 M − Magnesium chloride 10 mM − Spermine 2.5 mM − Calcium chloride 5 mM − Isopropanol 10% solution,
BioChemika 89263
Cacodylate pH 7.0; 0.05 M − Magnesium chloride 9 mM − Spermine 2.25 mM − Cobalt(III)-hexamine 1.8 mM − Spermidine 0.9 mM − PEG 400 5% solution,
BioChemika 89269
Cacodylate pH 6.5; 0.05 M − Magnesium chloride 10 mM − Spermine 2.5 mM − Copper sulfate 1 mM − Isopropanol 10% solution,
BioChemika 56961
Cacodylate pH 6.0; 0.05 M − Magnesium chloride 20 mM − Spermine 1.0 mM − Calcium chloride 2 mM − 1,5 Hexanediol 10% solution,
BioChemika 91373
HEPES pH 7.5; 0.05 M − Magnesium chloride 15 mM − Spermidine 1.0 mM − Dioxan 10% solution,
BioChemika 30193
Cacodylate pH 6.0; 0.05 M − Magnesium chloride 15 mM − Spermine 3.0 mM − PEG 400 10% solution,
BioChemika 67198
Cacodylate pH 6.5; 0.05 M − Spermine 2.5 mM − Calcium chloride 18 mM − 2-Propanol 9% solution,
BioChemika 97863
Cacodylate pH 6.5; 0.05 M − Spermine 2.0 mM − Cobalt(III)-hexamine 1.0 mM − Calcium chloride 80 mM solution,
BioChemika 55784
Cacodylate pH 6.5; 0.05 M − Magnesium chloride 5 mM − Cobalt(III)-hexamine 2.5 mM solution,
BioChemika 07312
Cacodylate pH 6.5; 0.05 M − Magnesium chloride 30 mM − Spermine 1.0 mM − Lithium sulfate 1.3 M solution,
BioChemika 19485
Cacodylate pH 6.0; 0.05 M − Calcium acetate 200 mM − Isopropanol 5% solution,
BioChemika 68446
Cacodylate pH 6.5; 0.05 M − Magnesium chloride 100 mM − Cobalt(III)-hexamine 1.0 mM − Ethanol 10% solution,
BioChemika 80215
Cacodylate pH 6.0; 0.05 M − Magnesium chloride 10 mM − Spermidine 2.5 mM − Sodium chloride 2.5 M solution,
BioChemika 19348
Cacodylate pH 6.5; 0.05 M − Magnesium chloride 10 mM − Sodium citrate 200 mM − Isopropanol 5% solution,
BioChemika 16829
Cacodylate pH 6.5; 0.05 M − Magnesium chloride 15 mM − Spermine 10.0 mM − Lithium sulfate 2.0 M solution,
BioChemika 44046
Cacodylate pH 6.5; 0.05 M − Magnesium chloride 20 mM − Spermine 1.0 mM − Ammonium sulfate 2.0 M solution,
BioChemika 73519
Cacodylate pH 6.5; 0.05 M − Magnesium chloride 10 mM − Spermine 1.5 mM − Ammonium sulfate 3.0 solution,
BioChemika 79853
HEPES pH 7.5; 0.05 M − Magnesium chloride 15 mM − Spermine 1.0 mM − Ammonium sulfate 1.0 M solution,
BioChemika 66017
Cacodylate pH 6.0; 0.05 M − Calcium acetate 200 mM − Sodium chloride 2.5 M solution,
BioChemika 18154
Cacodylate pH 6.0; 0.05 M − Calcium acetate 200 mM − Cobalt(III)-hexamine 1.0 mM − Lithium chloride 2.0 M solution,
BioChemika 42794
Cacodylate pH 6.5; 0.05 M − Magnesium chloride 15 mM − Spermidine 5.0 mM − Cobalt(III)-hexamine 1.0 mM − Sodium chloride 2.0 M solution,
BioChemika 40377
Cacodylate pH 6.5; 0.05 M − Magnesium chloride 200 mM − Sodium chloride 100 mM − PEG 1000 20% solution,
BioChemika 15873
TRIS pH 7.5; 0.05 M − Magnesium chloride 50 mM − Sodium tartrate 1.0 M solution,
BioChemika 42029
TRIS pH 7.5; 0.05 M − Magnesium chloride 200 mM − Sodium chloride 2.5 M solution,
BioChemika 44259
Cacodylate pH 6.0; 0.05 M − Magnesium chloride 200 mM − Potassium chloride 2.5 M solution,
BioChemika 41172
TRIS pH 8.0; 0.05 M – Magnesium chloride 200 mM – Ethanol 15% solution,
BioChemika 75771
Cacodylate pH 6.0; 0.05 M − Magnesium chloride 15 mM − Spermidine 5.0 mM − Lithium sulfate 2.0 M solution,
BioChemika 92213
Cacodylate pH 6.0; 0.05 M − Magnesium acetate 20 mM − Spermine 0.5 mM − Sodium chloride 100 mM − 2-Methyl-2,4-pentanediol 25% solution,
BioChemika 53245
Sodium succinate 0.05 M − Magnesium chloride 20 mM − Spermine 0.5 mM − Ammonium sulfate 3.0 solution,
BioChemika 43374
Cacodylate pH 6.5; 0.05 M − Cobalt(III)-hexamine 5 mM − Potassium chloride 2.5 M solution,
BioChemika 44401
Cacodylate pH 6.5; 0.05 M − Magnesium chloride 50 mM − Cobalt(III)-hexamine 2.0 mM − Lithium sulfate 1.5 M solution,
BioChemika 05556
Cacodylate pH 6.5; 0.05 M − Spermine 1.0 mM − Cobalt(III)-hexamine 2.0 mM − Calcium chloride 30 mM − Lithium chloride 2.0 M solution,
BioChemika 04359
Cacodylate pH 6.5; 0.05 M − Magnesium chloride 10 mM − Spermine 50 mM solution,
BioChemika 66953