product line
BioChemika
assay
≥99.0% (sum of enantiomers, HPLC)
optical activity
[α]20/D −20±1°, 5 hr, c = 4% in H2O
ign. residue
≤0.1% (as SO4)
loss
≤0.2% loss on drying, 20 °C (HV)
mp
85-88 °C
88-92 °C(lit.)
solubility
H2O: 0.1 g/mL, clear, colorless to light yellow
anion traces
chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤200 mg/kg
cation traces
As: ≤0.1 mg/kg
Ca: ≤100 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤20 mg/kg
Mn: ≤5 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg