Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
85067
Fluka
Sesame oil from
Sesamum indicum
Ph Eur
Synonym
Sesami oleum
CAS Number
8008-74-0
MDL number
MFCD00132243
EG/EC Number
232-370-6
Expand/Collapse All
Price and Availability
Properties
grade
Ph Eur
pharmacopeia
testing & handling conforms to Pharmacopeia
storage temp.
2-8°C
Safety
Safety Statements
24/25
WGK Germany
NWG
RTECS
VU3940000
F
8
Related Categories
...
Essential Biological Reagents
>
Lipids and Related Products
...
Lipids
>
Oils
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Similar Products
Related Categories
Page Options
Printer Friendly View
Ask A Scientist
Email Page
Last 5 Products Viewed
85067 (Fluka)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.