Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
92984
Fluka
Streptavidin B-Phycoerythrin
BioChemika
Expand/Collapse All
Price and Availability
Descriptions
Physical form
1 mg/ml in 0.1 M sodium phosphate, 0.1 M sodium chloride, 2 mM sodium azide, pH 7.5
Properties
product line
BioChemika
mol wt
M
r
~240000
fluorescence
λ
ex
542 nm; λ
em
575 nm in 0.1 M phosphate pH 7.0
suitability
corresponds for gel electrophoresis
storage temp.
2-8°C
Safety
Safety Statements
26-36/37/39-45
WGK Germany
-
F
10
Related Categories
...
Fluorescent Labels
>
Phycobili Proteins
...
Fluorescent Conjugates
>
Streptavidin/Avidin Conjugates
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Related Categories
Page Options
Printer Friendly View
Ask A Scientist
Email Page
Last 5 Products Viewed
92984 (Fluka)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.