Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Product No. Only)
Supplier X Reference
Substructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
BCR691
Fluka
Copper alloys (composition alloy)
BCR
®
certified Reference Material
Expand/Collapse All
Price and Availability
Descriptions
Packaging
set with 5 discs
Legal Information
BCR is a registered trademark of European Commission.
Properties
grade
BCR
®
certified Reference Material
Related Categories
...
C
>
CO - CZ
...
Matrix CRMs
>
Industrial Raw Materials
...
Industrial Raw Materials
>
Metals & Alloys
...
Matrix CRMs
>
Reactor Neutron Dosimetry
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Last 5 Products Viewed
BCR691 (Fluka)
en-us-footer US
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.