grade
ACS reagent
for determinations with dithizone
puriss. p.a.
assay
99.0-99.4% (GC)
contains
~1% ethanol as stabilizer
total impurities
free acid and chloride, complies
≤0.00001% free chlorine (Cl)
≤0.00005% free acid (as HCl)
≤0.0005% non-volatile matter
≤0.005% aldehydes and ketones (as CH3COCH3)
≤0.01% dichloromethane (GC)
≤0.01% tetrachloroethene (GC)
≤0.01% tetrachloromethane (GC)
≤0.01% water (Karl Fischer)
≤100% trichloroethene (GC)
0.6-1.0% ethanol (GC)
anion traces
chloride (Cl-): ≤1 mg/kg
cation traces
Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.05 mg/kg
Sn: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg
suitability
passes test for suitability of determ. w. dithizone