grade
puriss.
assay
98.0-100.5% (calc. for dried substance)
quality
meets analytical specification of Ph. Eur., BP, USP, FCC, E 530
filling volume
6.7-10 mL/g
total impurities
free alkali, in accordance
residual solvents, complies
soluble salts, in accordance
≤0.002% heavy metals (as Pb)
≤0.1% insoluble in HCl
≤0.1% insoluble in acetic acid
≤1% water-soluble matter
loss
≤5% loss on ignition
mp
2852 °C(lit.)
anion traces
chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤5000 mg/kg
cation traces
As: ≤2 mg/kg
Ca: ≤1%
Cu: ≤10 mg/kg
Fe: ≤200 mg/kg
Pb: ≤4 mg/kg
Zn: ≤10 mg/kg
suitability
passes test for appearance of solution