grade
puriss.
assay
99-101%
quality
meets analytical specification of Ph. Eur., BP, FCC, E511
total impurities
acidity or alkalinity, complies
residual solvents, complies
≤0.0001% Al
≤0.005% ammonium (NH4)
≤0.01% free alkali (as MgO)
≤0.02% free acid (as HCl)
51.0-55.0% water (Karl Fischer)
anion traces
bromide (Br-): ≤500 mg/kg
sulfate (SO42-): ≤100 mg/kg
cation traces
Al: ≤1 mg/kg
As: ≤2 mg/kg
Ba: ≤20 mg/kg
Ca: ≤1000 mg/kg
Cu: ≤10 mg/kg
Fe: ≤5 mg/kg
K: ≤500 mg/kg
Na: ≤5000 mg/kg
Pb: ≤4 mg/kg
Zn: ≤10 mg/kg
suitability
complies for appearance of solution