grade
ACS reagent
vapor density
2.07 (vs air)
vapor pressure
11.4 mmHg ( 20 °C)
assay
≥99.7%
autoignition temp.
800 °F
expl. lim.
16 %± , 92 °F
4 %± , 59 °F
total impurities
Cr2O72- reducers, passes test
MnO4- reducers, passes test
≤0.0004 meq/g Titr. base
≤0.01% acetic anhydride
evapn. residue
≤0.001%
color
≤10(APHA)
refractive index
n20/D 1.371(lit.)
bp
117-118 °C(lit.)
mp
16.2 °C(lit.)
density
1.049 g/mL at 25 °C(lit.)
anion traces
chloride (Cl-): ≤1 ppm
sulfate (SO42-): ≤1 ppm
cation traces
Fe: ≤0.2 ppm
heavy metals (as Pb): ≤0.5 ppm