grade
CHROMASOLV®
for HPLC
gradient grade
vapor density
1.11 (vs air)
vapor pressure
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)
assay
≥99.9%
autoignition temp.
725 °F
expl. lim.
36 %
total impurities
KMnO4 red. matter (as O), in accordance
≤0.00035% free alkali (as NH3)
≤0.0005% non-volatile matter
≤0.001% acetaldehyde
≤0.001% acetone (GC)
≤0.001% formaldehyde
≤0.02% water (Karl Fischer)
≤1 ppb fluorescence (quinine) at 254 nm
≤1 ppb fluorescence (quinine) at 365 nm
evapn. residue
<0.0005%
color
≤10(APHA)
transmittance
210 nm, ≥30%
220 nm, ≥50%
235 nm, ≥80%
260 nm, ≥98%
refractive index
n20/D 1.329(lit.)
bp
64.7 °C(lit.)
mp
−98 °C(lit.)
density
0.791 g/mL at 25 °C(lit.)
abs.
/210 nm 0.500
/220 nm 0.300
HPLC-gradient/230 nm ≤2 mAU
/235 nm 0.100
HPLC-gradient/254 nm ≤5 mAU
/260 nm 0.010
absorption/ in accordance
suitability
in accordance for appearance
in accordance for miscibility (with H2O)
in accordance for reaction against H2SO4