![]() |
Synonym
ACh
Molecular Formula
(CH3)3N+CH2CH2OCOCH3Cl-
Molecular Weight
181.66
CAS Number
Beilstein Registry Number
3571875
MDL number
PubChem Substance ID
EG/EC Number
200-468-8
References
Reference
Perry, E., et al., Acetylcholine in mind: a neurotransmitter correlate of consciousness?. Trends Neurosci. 22, 273-280, (1999)
Israel, M., and Dunant, Y., Acetylcholine release and the cholinergic genomic locus. Mol. Neurobiol. 16, 1-20, (1998)
Pepeu, G., and Blandina, P., The acetylcholine, GABA, glutamate triangle in the rat forebrain. J. Physiol. (Paris) 92, 351-355, (1998)
Wessler, I., et al., The cholinergic 'pitfall': acetylcholine, a universal cell molecule in biological systems, including humans. Clin. Exp. Pharmacol. Physiol. 26, 198-205, (1999)
Merck
Merck 13,89
Beilstein
Beil. 4,IV,1446
reference
Aldrich MSDS 1, 19:B / Corp MSDS 1 (1), 35:D / FT-IR 2 (1), 1130:A / FT-IR 1 (1), 678:C / FT-NMR 1 (1), 1078:A / IR-Spectra (3), 396:E / IR-Spectra (2), 350:H / RegBook 1 (1), 773:H / Sax 6, 96 / Sigma FT-IR 1 (2), 1:C / Structure Index 1, 120:B:7
| ||||||||