![]() |
Synonym
(2
Molecular Formula
(CH3)3N(Cl)CH2CH2OH
Molecular Weight
139.62
CAS Number
Beilstein Registry Number
3563126
MDL number
PubChem Substance ID
EG/EC Number
200-655-4
References
Reference
Jimenez, B., et al., Occurrence, co-occurence and topographic distribution of choline acetyl transferase, met-enkephalin and neurotensin in the stellate ganglion of the cat. Synapse 43, 163, (2002)
Merck
Merck 13,2226
Beilstein
Beil. 4,IV,1443
reference
Aldrich MSDS 1, 468:B / Corp MSDS 1 (1), 876:C / FT-IR 2 (1), 621:C / FT-IR 1 (1), 395:D / IR-Spectra (3), 231:E / IR-Spectra (2), 206:F / NMR-Reference 2 (1), 347:D / RegBook 1 (1), 421:J / Sax 6, 788 / Sigma FT-IR 1 (2), 9:A / Structure Index 1, 61:B:9
| ||||||||