Application
Not suitable for use as attachment factor or 3-D matrix.
General description
Collagen terminology using the Bornstein and Traub designation originates from the reference; Bornstein, P. and Traub, W. The Proteins, (1979) 4, 411-605
Collagen is classified into a number of structurally and genetically distinct types. We use the nomenclature proposed by Bornstein and Traub.1 Do not confuse Sigma type designations with recognized collagen classification types.
Other Notes
Bornstein and Traub Type I
Preparation Note
Prepared by the method of Einbinder, J. and Schubert, M., J. Biol. Chem., 188, 335 (1951).