![]() |
Synonym
1,2,3
Glycerin
Molecular Formula
HOCH2CH(OH)CH2OH
Molecular Weight
92.09
CAS Number
Beilstein Registry Number
635685
MDL number
PubChem Substance ID
EG/EC Number
200-289-5
Descriptions
Application
Glycerol is used both in sample preparation and gel formation for polyacrylamide gel electrophoresis. Glycerol (5-10%) increases the density of a sample so that the sample will layer at the bottom of a gel’s sample well. Glycerol is also used to aid in casting gradient gels and as a protein stabilizer and storage buffer component.
Packaging
1 gal in glass btl
1 L in glass btl
20 L in steel drum
500 mL in glass btl
Legal Information
ReagentPlus is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.
References
Reference
, Andrews, A.T. Electrophoresis: Theory, Techniques, and Biochemical and Clinical Applications 2nd ed., New York , (1988), 252
, Link, A.J. ed., 2-D Proteome Analysis Protocols. Methods Mol. Biol. Totowa, NJ , (1999) 112
Merck
Merck 13,4497
Beilstein
Beil. 1,IV,2751
reference
Aldrich MSDS 1, 993:A / Corp MSDS 1 (1), 1782:D / FT-IR 2 (1), 265:B / FT-IR 1 (1), 183:A / FT-NMR 1 (1), 283:C / IR-Spectra (3), 112:A / IR-Spectra (2), 103:G / NMR-Reference 2 (1), 171:B / RegBook 1 (1), 185:A / Sax 6, 1479 / Sigma FT-IR 1 (1), 1340:B / Structure Index 1, 24:C:7 / Vapor Phase 3, 262:A
| ||||||||