![]() |
Synonym
1,2,3,5/4,6
i
meso
Molecular Formula
C6H12O6
Molecular Weight
180.16
CAS Number
Beilstein Registry Number
1907329
MDL number
PubChem Substance ID
EG/EC Number
201-781-2
References
Reference
Wang, W.T., et al., Analysis of inositol by high-performance liquid chromatography. Anal. Biochem. 188, 432, (1990)
Ferguson, M.A., Williams, A.F., Cell-surface anchoring of proteins via glycosyl-phosphatidylinositol structures. Annu. Rev. Biochem. 57, 285, (1988)
Merck
Merck 13,5001
Beilstein
Beil. 6,IV,7919
reference
Corp MSDS 1 (1), 1985:B / FT-IR 2 (1), 248:D / FT-IR 1 (1), 173:C / FT-NMR 1 (1), 266:A / IR-Spectra (3), 107:A / IR-Spectra (2), 99:D / NMR-Reference 2 (1), 164:A / RegBook 1 (1), 173:I / Sigma FT-IR 1 (1), 1089:C / Structure Index 1, 22:C:9
| ||||||||