Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
M8662
Sigma-Aldrich
Mineral oil
PCR Reagent
CAS Number
8042-47-5
MDL number
MFCD00131611
EG/EC Number
232-455-8
Expand/Collapse All
Price and Availability
Properties
grade
PCR Reagent
packaging
vial of 1.5 mL
refractive index
n
20/D
1.467(lit.)
density
0.84 g/mL at 25 °C(lit.)
foreign activity
DNase, RNase, protease, none detected
References
reference
Corp MSDS
1
(2), 2468:D /
FT-IR
2
(1), 12:C /
FT-IR
2
(1), 12:B /
FT-IR
1
(1), 7:C /
FT-NMR
1
(1), 9:C /
FT-NMR
1
(1), 10:A /
NMR-Reference
2
(1), 13:D /
Sax
6
, 1949 /
Sigma FT-IR
1
(2), 758:D
Safety
WGK Germany
1
RTECS
PY8047000
Related Categories
...
PCR/Amplification
>
PCR Reaction Components
...
Essential Molecular Biology Reagents
>
PCR Reagents
Scroll Up
Scroll Down
Related Information
M8662 - Datasheet (48kb)
PCR Prod Guide (228kb)
PCR Prod Guide (318kb)
MSDS
Certificate of Analysis
More Information
Links
Similar Products
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Email Page
Last 5 Products Viewed
M8662 (Sigma-Aldrich)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.